Tisk…
Anti-Stage Specific Embryonic Antigen 4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 8H218]
Dodavatel: United States Biological
USBIS6610-11AEA 21450 CZK
USBIS6610-11A
Anti-Stage Specific Embryonic Antigen 4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 8H218]
Protilátky
Do not freeze!
Type: Primary
Antigen: Stage Specific Embryonic Antigen 4
Clonality: Monoclonal
Clone: 8H218
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Human, Mouse