Tisk…

Anti-Transforming Growth Factor beta Receptor III Goat Polyclonal Antibody (Carboxyfluorescein)

Dodavatel: United States Biological
USBIT8250-50FEA 15200 CZK
USBIT8250-50F
Anti-Transforming Growth Factor beta Receptor III Goat Polyclonal Antibody (Carboxyfluorescein)
Protilátky
Type: Primary
Antigen: Transforming Growth Factor beta Receptor III
Clonality: polyclonal
Clone:
Conjugation: Carboxyfluorescein
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR