Tisk…

Anti-TGFBR3 Rabbit Polyclonal Antibody

Dodavatel: United States Biological
USBIT8250-50HEA 16010 CZK
USBIT8250-50H
Anti-TGFBR3 Rabbit Polyclonal Antibody
Protilátky
Recommended dilution: Immunochemistry: 1:50

Type: Primary
Antigen: TGFBR3 (transforming growth factor, beta receptor III)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR