Tisk…

Anti-TGFBI Rabbit Monoclonal Antibody [clone: 11C1085]

Dodavatel: United States Biological
USBIT8252-91HEA 19030 CZK
USBIT8252-91H
Anti-TGFBI Rabbit Monoclonal Antibody [clone: 11C1085]
Protilátky
Recommended dilution: Immunofluorescence: 1:800; Western Blot: 1:1000; Immunoprecipitation: 1:100

Type: Primary
Antigen: TGFBI (transforming growth factor, beta-induced, 68kDa)
Clonality: monoclonal
Clone: 11C1085
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR