Tisk…

Anti-Transforming Growth Factor beta Induced Goat Polyclonal Antibody

Dodavatel: United States Biological
USBIT8252-92EA 15540 CZK
USBIT8252-92
Anti-Transforming Growth Factor beta Induced Goat Polyclonal Antibody
Protilátky
Recommended dilution: ELISA: 1: 32000; Western Blot: 02-0.5 ?g/ml

Type: Primary
Antigen: Transforming Growth Factor beta Induced
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR