Tisk…

Anti-TGM4 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Dodavatel: United States Biological
USBIT8260-20B-APCEA 24020 CZK
USBIT8260-20B-APC
Anti-TGM4 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Protilátky
Recommended dilution: FLISA: 1:1000; Flow Cytometry: 1:10-1:50; Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: TGM4 (transglutaminase 4)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR