Tisk…

Anti-IgG Donkey Polyclonal Antibody (AP (Alkaline Phosphatase))

Dodavatel: Immunoreagents
IMMRIR1202EA 4940 CZK
IMMRIR1202
Anti-IgG Donkey Polyclonal Antibody (AP (Alkaline Phosphatase))
Protilátky
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Rabbit
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR