Tisk…

Anti-Hemoglobin Goat Polyclonal Antibody

Dodavatel: Immunoreagents
IMMRIR1480EA 7870 CZK
IMMRIR1480
Anti-Hemoglobin Goat Polyclonal Antibody
Protilátky
Type: Primary
Antigen: Hemoglobin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR