Tisk…

Anti-kappa light chain Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Immunoreagents
IMMRIR1503EA 8320 CZK
IMMRIR1503
Anti-kappa light chain Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Type: Secondary
Antigen: kappa light chain
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR