Tisk…

Goat Anti-Rabbit IgG Polyclonal Antibody (DyLight® 633)

IMMRIR2169EA 3440 CZK
IMMRIR2169
Goat Anti-Rabbit IgG Polyclonal Antibody (DyLight® 633)
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 633
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR