Tisk…

Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Immunoreagents
IMMRIR2219EA 1500 CZK
IMMRIR2219
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR