Tisk…
Eppendorf Tubes® 15/50 ml, with screw cap
Dodavatel: Eppendorf
525-1054EA 4760 CZK
525-1054 525-1053 525-1052 525-1051 525-1050 525-1049
Eppendorf Tubes® 15/50 ml, with screw cap
Zkumavky Zkumavky centrifugační
PP, with HDPE scew cap.
  • High quality material and precision molding ensures a high tightness and minimises sample loss
  • Ergonomic handling (easy opening/closing) and one-handed operation
  • Reliable and safe labelling
  • Each individual lot is checked and certified for quality and purity
Safe design and easy handling ensure best sample processing. They represent the optimal solution for cell culture and cell biology applications as well as for sample preparation protocols in microbiology and molecular biology laboratories.
DNA LoBind tubes
DNA LoBind tubes maximise sample recovery of nucleic acids by significantly reducing sample to surface binding. They represent the ideal solution for sample preparation and storage of samples containing low amounts of DNA.
Sterile tubes
Pyrogen-, DNase-, RNase-, human and bacterial DNA-free.