Tisk…
Food Stamp /Clean Stamp
SCIL1000043EA 9690 CZK
SCIL1000043
Food Stamp /Clean Stamp
Food Stamp /Clean Stamp -fFor hygiene inspection on foods and for environmental surface sampling. Food Stamp Rodac (Replicate Organism Detection and Counting) plates are a simple-to-use bacteriological method to check critical control points (CCP's) within the HACCP (hazardous analytical critical control points) for the microbiological hygiene of surfaces. Food Stamps are designated for hygiene control of viable bacteria on foodstuffs and working areas. It is a simple-to-use bacteriological method which indicates the presence of bacteria.
  • No special instruments or equipment required
  • Time saving as no time for preparation is needed
  • Simple and easy test leads to reliable results
  • Unique products utilizing a chromogenic reaction give more distinctive color development for easy reading
  • Agar inside the container is firmly attached to prevent the disintegration
  • Compact and easy to handle
  • Portable incubator: IncuBox which is specially designed for Food Stamps (up to 30 plates)