Tisk…

Anti-IgG Antibody (Biotin)

Dodavatel: Boster
BSBTBA1006-0.5EA 0 CZK
BSBTBA1006-0.5
Anti-IgG Antibody (Biotin)
Protilátky
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: Biotin
Epitope:
Host:
Isotype:
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR