Tisk…

Anti-IgG Antibody (Cy3®)

Dodavatel: Boster
BSBTBA1035EA 6740 CZK
BSBTBA1035
Anti-IgG Antibody (Cy3®)
Protilátky
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: Cy3
Epitope:
Host:
Isotype:
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR