Tisk…

Anti-Protein A Antibody (5nm colloidal Gold)

Dodavatel: Boster
BSBTGA1053EA 6230 CZK
BSBTGA1053
Anti-Protein A Antibody (5nm colloidal Gold)
Protilátky
Type: Tag
Antigen: Protein A
Clonality:
Clone:
Conjugation: 5nm colloidal Gold
Epitope:
Host:
Isotype:
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR