Tisk…

Anti-IgG Antibody (FITC (Fluorescein Isothiocyanate))

Dodavatel: Boster
BSBTBA1101-0.5EA 0 CZK
BSBTBA1101-0.5
Anti-IgG Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host:
Isotype:
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR