Tisk…

Anti-GGPS1 Mouse Monoclonal Antibody [clone: 2E7]

Dodavatel: OriGene
ORIGCF501729EA 21120 CZK
ORIGCF501729
Anti-GGPS1 Mouse Monoclonal Antibody [clone: 2E7]
Protilátky
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: GGPS1
Clonality: Monoclonal
Clone: 2E7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR