Tisk…

Anti-LMAN1 Mouse Monoclonal Antibody [clone: 2B10]

Dodavatel: OriGene
ORIGCF502110EA 21120 CZK
ORIGCF502110
Anti-LMAN1 Mouse Monoclonal Antibody [clone: 2B10]
Protilátky
Recommended Dilutions: Western Blot: 1:200-500; Immunofluorescence: 1:100

Type: Primary
Antigen: LMAN1
Clonality: Monoclonal
Clone: 2B10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR