Tisk…

Anti-MYD88 Mouse Monoclonal Antibody [clone: 1B8]

Dodavatel: OriGene
ORIGCF502118EA 21710 CZK
ORIGCF502118
Anti-MYD88 Mouse Monoclonal Antibody [clone: 1B8]
Protilátky
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MYD88
Clonality: Monoclonal
Clone: 1B8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR