Tisk…

Anti-DNAJA2 Mouse Monoclonal Antibody [clone: 1G9]

Dodavatel: OriGene
ORIGCF502126EA 21120 CZK
ORIGCF502126
Anti-DNAJA2 Mouse Monoclonal Antibody [clone: 1G9]
Protilátky
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: DNAJA2
Clonality: Monoclonal
Clone: 1G9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR