Tisk…

Anti-STAT5A Mouse Monoclonal Antibody [clone: 6D6]

Dodavatel: OriGene
ORIGCF502739EA 21120 CZK
ORIGCF502739
Anti-STAT5A Mouse Monoclonal Antibody [clone: 6D6]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: STAT5A
Clonality: Monoclonal
Clone: 6D6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR