Tisk…

Anti-ADH1B Mouse Monoclonal Antibody [clone: 5D7]

Dodavatel: OriGene
ORIGCF502778EA 21120 CZK
ORIGCF502778
Anti-ADH1B Mouse Monoclonal Antibody [clone: 5D7]
Protilátky
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ADH1B
Clonality: Monoclonal
Clone: 5D7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Monkey
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR