Tisk…

Anti-NME4 Mouse Monoclonal Antibody [clone: 2G1]

Dodavatel: OriGene
ORIGCF501116EA 21120 CZK
ORIGCF501116
Anti-NME4 Mouse Monoclonal Antibody [clone: 2G1]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: NME4
Clonality: Monoclonal
Clone: 2G1
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR