Tisk…

Anti-NME4 Mouse Monoclonal Antibody [clone: 4F10]

Dodavatel: OriGene
ORIGCF501141EA 21120 CZK
ORIGCF501141
Anti-NME4 Mouse Monoclonal Antibody [clone: 4F10]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: NME4
Clonality: Monoclonal
Clone: 4F10
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR