Tisk…

Anti-NEK11 Mouse Monoclonal Antibody [clone: 5D9]

Dodavatel: OriGene
ORIGCF501765EA 21710 CZK
ORIGCF501765
Anti-NEK11 Mouse Monoclonal Antibody [clone: 5D9]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: NEK11
Clonality: Monoclonal
Clone: 5D9
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR