Tisk…

Anti-TCP11L2 Mouse Monoclonal Antibody [clone: 3E4]

Dodavatel: OriGene
ORIGCF501806EA 21710 CZK
ORIGCF501806
Anti-TCP11L2 Mouse Monoclonal Antibody [clone: 3E4]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: TCP11L2
Clonality: Monoclonal
Clone: 3E4
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR