Tisk…

Anti-PPM1B Mouse Monoclonal Antibody [clone: 3A10]

Dodavatel: OriGene
ORIGCF502862EA 21120 CZK
ORIGCF502862
Anti-PPM1B Mouse Monoclonal Antibody [clone: 3A10]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: PPM1B
Clonality: Monoclonal
Clone: 3A10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR