Tisk…

Anti-ERCC1 Mouse Monoclonal Antibody [clone: 1C6]

Dodavatel: OriGene
ORIGCF501169EA 21120 CZK
ORIGCF501169
Anti-ERCC1 Mouse Monoclonal Antibody [clone: 1C6]
Protilátky
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100; Flow Cytometry: 1:100; Immunochemistry: 1:50-1:150

Type: Primary
Antigen: ERCC1
Clonality: Monoclonal
Clone: 1C6
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR