Tisk…

Anti-ERCC1 Mouse Monoclonal Antibody [clone: 1H1]

Dodavatel: OriGene
ORIGCF501184EA 21120 CZK
ORIGCF501184
Anti-ERCC1 Mouse Monoclonal Antibody [clone: 1H1]
Protilátky
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ERCC1
Clonality: Monoclonal
Clone: 1H1
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR