Tisk…

Anti-BSG Mouse Monoclonal Antibody [clone: 5D8]

Dodavatel: OriGene
ORIGCF501190EA 21120 CZK
ORIGCF501190
Anti-BSG Mouse Monoclonal Antibody [clone: 5D8]
Protilátky
Recommended Dilutions: Western Blot: 1:200-1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: BSG
Clonality: Monoclonal
Clone: 5D8
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR