Tisk…

Anti-BSG Mouse Monoclonal Antibody [clone: 5E1]

Dodavatel: OriGene
ORIGCF501160EA 21120 CZK
ORIGCF501160
Anti-BSG Mouse Monoclonal Antibody [clone: 5E1]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: BSG
Clonality: Monoclonal
Clone: 5E1
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR