Tisk…

Anti-KRT18 Mouse Monoclonal Antibody [clone: 1E1]

Dodavatel: OriGene
ORIGCF500015EA 21120 CZK
ORIGCF500015
Anti-KRT18 Mouse Monoclonal Antibody [clone: 1E1]
Protilátky
Recommended Dilutions:Western Blot: 1:500-1000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: KRT18
Clonality: Monoclonal
Clone: 1E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR