Tisk…

Anti-KRT18 Mouse Monoclonal Antibody [clone: 5E8]

Dodavatel: OriGene
ORIGCF500016EA 21120 CZK
ORIGCF500016
Anti-KRT18 Mouse Monoclonal Antibody [clone: 5E8]
Protilátky
Recommended Dilutions:Western Blot: 1:1000; Immunochemistry: 1:50

Type: Primary
Antigen: KRT18
Clonality: Monoclonal
Clone: 5E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR