Tisk…

Anti-KRT8 Mouse Monoclonal Antibody [clone: 2E4]

Dodavatel: OriGene
ORIGCF500022EA 21120 CZK
ORIGCF500022
Anti-KRT8 Mouse Monoclonal Antibody [clone: 2E4]
Protilátky
Recommended Dilutions:Western Blot: 1:500-1000; Immunochemistry: 1:50; Immunofluorescence: 1:50-100

Type: Primary
Antigen: KRT8
Clonality: Monoclonal
Clone: 2E4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR