Tisk…

Anti-Nog Mouse Monoclonal Antibody [clone: 4C12]

Dodavatel: OriGene
ORIGCF500028EA 21120 CZK
ORIGCF500028
Anti-Nog Mouse Monoclonal Antibody [clone: 4C12]
Protilátky
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: Nog
Clonality: Monoclonal
Clone: 4C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR