Tisk…

Anti-NTF3 Mouse Monoclonal Antibody [clone: 2D1]

Dodavatel: OriGene
ORIGCF500031EA 21120 CZK
ORIGCF500031
Anti-NTF3 Mouse Monoclonal Antibody [clone: 2D1]
Protilátky
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: NTF3
Clonality: Monoclonal
Clone: 2D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR