Tisk…

Anti-NTF4 Mouse Monoclonal Antibody [clone: 2E8]

Dodavatel: OriGene
ORIGCF500034EA 21120 CZK
ORIGCF500034
Anti-NTF4 Mouse Monoclonal Antibody [clone: 2E8]
Protilátky
Recommended Dilutions: Western Blot: 1:5000; Immunochemistry: 1:50; Immunofluorescence: 1:50

Type: Primary
Antigen: NTF4
Clonality: Monoclonal
Clone: 2E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR