Tisk…

Anti-CDCP1 Mouse Monoclonal Antibody [clone: 2B8]

Dodavatel: OriGene
ORIGCF502171EA 21120 CZK
ORIGCF502171
Anti-CDCP1 Mouse Monoclonal Antibody [clone: 2B8]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Flow Cytometry: 1:100

Type: Primary
Antigen: CDCP1
Clonality: Monoclonal
Clone: 2B8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR