Tisk…

Anti-NHEJ1 Mouse Monoclonal Antibody [clone: 1G5]

Dodavatel: OriGene
ORIGCF502176EA 21710 CZK
ORIGCF502176
Anti-NHEJ1 Mouse Monoclonal Antibody [clone: 1G5]
Protilátky
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: NHEJ1
Clonality: Monoclonal
Clone: 1G5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR