Tisk…

Anti-MAPK7 Mouse Monoclonal Antibody [clone: 4D6]

Dodavatel: OriGene
ORIGCF502188EA 21120 CZK
ORIGCF502188
Anti-MAPK7 Mouse Monoclonal Antibody [clone: 4D6]
Protilátky
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MAPK7
Clonality: Monoclonal
Clone: 4D6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR