Tisk…

Anti-CRYAB Mouse Monoclonal Antibody [clone: 6D11]

Dodavatel: OriGene
ORIGCF500583EA 21120 CZK
ORIGCF500583
Anti-CRYAB Mouse Monoclonal Antibody [clone: 6D11]
Protilátky
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:50; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: CRYAB
Clonality: Monoclonal
Clone: 6D11
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR