Tisk…

Anti-POR Mouse Monoclonal Antibody [clone: 3H10]

Dodavatel: OriGene
ORIGCF500572EA 21120 CZK
ORIGCF500572
Anti-POR Mouse Monoclonal Antibody [clone: 3H10]
Protilátky
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: POR
Clonality: Monoclonal
Clone: 3H10
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR