Tisk…

Anti-PRKG1 Mouse Monoclonal Antibody [clone: 9A4]

Dodavatel: OriGene
ORIGCF501208EA 21120 CZK
ORIGCF501208
Anti-PRKG1 Mouse Monoclonal Antibody [clone: 9A4]
Protilátky
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PRKG1
Clonality: Monoclonal
Clone: 9A4
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR