Tisk…

Anti-PFN1 Mouse Monoclonal Antibody [clone: 1D5]

Dodavatel: OriGene
ORIGCF501211EA 21710 CZK
ORIGCF501211
Anti-PFN1 Mouse Monoclonal Antibody [clone: 1D5]
Protilátky
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PFN1
Clonality: Monoclonal
Clone: 1D5
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Dog, Human, Monkey, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR