Tisk…

Anti-ACAT2 Mouse Monoclonal Antibody [clone: 3C9]

Dodavatel: OriGene
ORIGCF501223EA 21120 CZK
ORIGCF501223
Anti-ACAT2 Mouse Monoclonal Antibody [clone: 3C9]
Protilátky
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ACAT2
Clonality: Monoclonal
Clone: 3C9
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2b
Reactivity: Dog, Human, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR