Tisk…

Anti-H6PD Mouse Monoclonal Antibody [clone: 3H5]

Dodavatel: OriGene
ORIGCF501242EA 21120 CZK
ORIGCF501242
Anti-H6PD Mouse Monoclonal Antibody [clone: 3H5]
Protilátky
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: H6PD
Clonality: Monoclonal
Clone: 3H5
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR