Tisk…

Anti-DTNA Mouse Monoclonal Antibody [clone: 1G1]

Dodavatel: OriGene
ORIGCF502223EA 21120 CZK
ORIGCF502223
Anti-DTNA Mouse Monoclonal Antibody [clone: 1G1]
Protilátky
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100

Type: Primary
Antigen: DTNA
Clonality: Monoclonal
Clone: 1G1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR