Tisk…

Anti-BIRC5 Mouse Monoclonal Antibody [clone: 6H5]

Dodavatel: OriGene
ORIGCF502235EA 21120 CZK
ORIGCF502235
Anti-BIRC5 Mouse Monoclonal Antibody [clone: 6H5]
Protilátky
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100

Type: Primary
Antigen: BIRC5
Clonality: Monoclonal
Clone: 6H5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR