Tisk…

Anti-POR Mouse Monoclonal Antibody [clone: 4G2]

Dodavatel: OriGene
ORIGCF500591EA 21120 CZK
ORIGCF500591
Anti-POR Mouse Monoclonal Antibody [clone: 4G2]
Protilátky
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Flow Cytometry: 1:100

Type: Primary
Antigen: POR
Clonality: Monoclonal
Clone: 4G2
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR